Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCTGTATCCCAGTTTCCCAGCA[A/C]TGGACATTATGAAAGACCAGTGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608639 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHA8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PLEKHA8 - pleckstrin homology domain containing A8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197026.1 | Intron | NP_001183955.1 | ||||
NM_001197027.1 | Intron | NP_001183956.1 | ||||
NM_032639.3 | Intron | NP_116028.1 | ||||
XM_005249897.2 | Intron | XP_005249954.1 | ||||
XM_011515591.2 | Intron | XP_011513893.1 | ||||
XM_011515592.2 | Intron | XP_011513894.1 | ||||
XM_011515594.1 | Intron | XP_011513896.1 | ||||
XM_011515595.1 | Intron | XP_011513897.1 | ||||
XM_011515596.2 | Intron | XP_011513898.1 | ||||
XM_017012744.1 | Intron | XP_016868233.1 | ||||
XM_017012745.1 | Intron | XP_016868234.1 | ||||
XM_017012746.1 | Intron | XP_016868235.1 | ||||
XM_017012747.1 | Intron | XP_016868236.1 | ||||
XM_017012748.1 | Intron | XP_016868237.1 | ||||
XM_017012749.1 | Intron | XP_016868238.1 |
Set Membership: |
HapMap |