Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAGTGTTCTTGCAATAGTAGCAC[C/T]AGCAGATTTTGTAAGACAATTTCTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612087 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLEC2A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CLEC2A - C-type lectin domain family 2 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130711.1 | 878 | Intron | NP_001124183.1 | |||
NM_207375.2 | 878 | Intron | NP_997258.1 | |||
XM_011520654.2 | 878 | Intron | XP_011518956.1 | |||
XM_011520656.2 | 878 | Intron | XP_011518958.1 | |||
XM_011520657.2 | 878 | UTR 3 | XP_011518959.1 |
KLRF2 - killer cell lectin like receptor F2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190765.1 | 878 | Intron | NP_001177694.1 |
Set Membership: |
HapMap |