Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATTATAATGATGTCTTTATGTTC[A/G]TCTTAGGAGGTGATAGTACTGATTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600207 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HPCAL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HPCAL1 - hippocalcin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258357.1 | Intron | NP_001245286.1 | ||||
NM_001258358.1 | Intron | NP_001245287.1 | ||||
NM_001258359.1 | Intron | NP_001245288.1 | ||||
NM_002149.3 | Intron | NP_002140.2 | ||||
NM_134421.2 | Intron | NP_602293.1 | ||||
XM_005246160.1 | Intron | XP_005246217.1 | ||||
XM_005246161.1 | Intron | XP_005246218.1 | ||||
XM_005246162.1 | Intron | XP_005246219.1 | ||||
XM_005246163.1 | Intron | XP_005246220.1 | ||||
XM_011510346.2 | Intron | XP_011508648.1 | ||||
XM_011510347.1 | Intron | XP_011508649.1 | ||||
XM_011510348.1 | Intron | XP_011508650.1 | ||||
XM_011510349.1 | Intron | XP_011508651.1 | ||||
XM_017003950.1 | Intron | XP_016859439.1 | ||||
XM_017003951.1 | Intron | XP_016859440.1 | ||||
XM_017003952.1 | Intron | XP_016859441.1 | ||||
XM_017003953.1 | Intron | XP_016859442.1 |