Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGCAGGGTGTTGGGATTTCCATA[A/C]AGTCTGACCGTGCAGTTGCTCAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602971 | ||||||||||||||||||||
Literature Links: |
GLTSCR1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLTSCR1L - GLTSCR1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBCC - tubulin folding cofactor C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003192.2 | 727 | Silent Mutation | CTG,CTT | L,L 218 | NP_003183.1 |