Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCCAGTCCTCCTGAGGGCCCAC[A/T]TTTTTCTGGGATCACCTGTTTTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611332 | ||||||||||||||||||||
Literature Links: |
DNAJB6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAJB6 - DnaJ heat shock protein family (Hsp40) member B6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005494.2 | Intron | NP_005485.1 | ||||
NM_058246.3 | Intron | NP_490647.1 | ||||
XM_005249515.2 | Intron | XP_005249572.1 | ||||
XM_005249516.2 | Intron | XP_005249573.1 | ||||
XM_006715823.1 | Intron | XP_006715886.1 | ||||
XM_011515704.1 | Intron | XP_011514006.1 | ||||
XM_017011633.1 | Intron | XP_016867122.1 |