Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAAATATGGCTTATTGTTTTGTT[A/G]ATGAATTTAAGAAACTTCTAAAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 172439 MIM: 602122 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAICS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PAICS - phosphoribosylaminoimidazole carboxylase; phosphoribosylaminoimidazolesuccinocarboxamide synthase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRP72 - signal recognition particle 72 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267722.1 | Intron | NP_001254651.1 | ||||
NM_006947.3 | Intron | NP_008878.3 | ||||
XM_005265765.4 | Intron | XP_005265822.3 |