Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTCTTTTGACTTAGGAAAAATA[C/T]GGTTTCTAATGCGAAATGTTGGTGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHG1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PLEKHG1 - pleckstrin homology and RhoGEF domain containing G1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005267064.3 | Intron | XP_005267121.1 | ||||
XM_005267065.3 | Intron | XP_005267122.1 | ||||
XM_005267066.2 | Intron | XP_005267123.1 | ||||
XM_006715521.1 | Intron | XP_006715584.1 | ||||
XM_006715522.3 | Intron | XP_006715585.1 | ||||
XM_011535981.2 | Intron | XP_011534283.1 | ||||
XM_011535982.2 | Intron | XP_011534284.1 | ||||
XM_011535983.2 | Intron | XP_011534285.1 | ||||
XM_017011102.1 | Intron | XP_016866591.1 |