Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTGGATGAGGCCTCTCCTTTTA[C/T]ATGCAGATCCTGGATTTCCACAGAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613242 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R14C PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PPP1R14C - protein phosphatase 1 regulatory inhibitor subunit 14C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030949.2 | Intron | NP_112211.1 | ||||
XM_005267152.4 | Intron | XP_005267209.1 |
Set Membership: |
HapMap |