Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGAACAAAATAATGTAGATTAT[A/T]AACACATTTTAACACATTCACATGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603128 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIM1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SIM1 - single-minded family bHLH transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005068.2 | 6014 | Intron | NP_005059.2 | |||
XM_005267100.2 | 6014 | UTR 3 | XP_005267157.1 | |||
XM_011536072.2 | 6014 | Intron | XP_011534374.1 | |||
XM_017011197.1 | 6014 | UTR 3 | XP_016866686.1 |
Set Membership: |
HapMap |