Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGTGCTTCTGCAATACTGTCATT[C/T]GAAGGTGGGGCATCAGGATCAGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC285638 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC285638 - uncharacterized LOC285638 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PJA2 - praja ring finger ubiquitin ligase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014819.4 | 2396 | Silent Mutation | TCA,TCG | S,S 700 | NP_055634.3 | |
XM_017010099.1 | 2396 | Silent Mutation | TCA,TCG | S,S 700 | XP_016865588.1 |