Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTAAACTTCTTACTTAAGACTGT[A/G]TGGAAATGTAGAGTGAGACAGAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603969 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNFSF13B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TNFSF13B - tumor necrosis factor superfamily member 13b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145645.2 | Intron | NP_001139117.1 | ||||
NM_006573.4 | Intron | NP_006564.1 |