Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTCAAAGCCACATTGGACTCATT[G/T]TTTCTGCTACCTTTATAAACCTTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602376 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFNAR2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IFNAR2 - interferon alpha and beta receptor subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000874.4 | Intron | NP_000865.2 | ||||
NM_001289125.1 | Intron | NP_001276054.1 | ||||
NM_001289126.1 | Intron | NP_001276055.1 | ||||
NM_001289128.1 | Intron | NP_001276057.1 | ||||
NM_207584.2 | Intron | NP_997467.1 | ||||
NM_207585.2 | Intron | NP_997468.1 |
Set Membership: |
HapMap |