Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGAATAAAAGTAGGGCTAAACAG[A/G]CAAGATGCAAATACATTTTCAAGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610450 MIM: 608789 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYPD1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
LYPD1 - LY6/PLAUR domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NCKAP5 - NCK associated protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207363.2 | Intron | NP_997246.2 | ||||
NM_207481.3 | Intron | NP_997364.3 | ||||
XM_005263659.3 | Intron | XP_005263716.1 | ||||
XM_005263660.3 | Intron | XP_005263717.1 | ||||
XM_011511097.2 | Intron | XP_011509399.1 | ||||
XM_011511098.2 | Intron | XP_011509400.1 | ||||
XM_011511099.2 | Intron | XP_011509401.1 | ||||
XM_011511100.2 | Intron | XP_011509402.1 | ||||
XM_011511101.1 | Intron | XP_011509403.1 | ||||
XM_011511102.2 | Intron | XP_011509404.1 | ||||
XM_011511103.2 | Intron | XP_011509405.1 | ||||
XM_011511104.1 | Intron | XP_011509406.1 | ||||
XM_011511105.2 | Intron | XP_011509407.1 | ||||
XM_017003974.1 | Intron | XP_016859463.1 | ||||
XM_017003975.1 | Intron | XP_016859464.1 | ||||
XM_017003976.1 | Intron | XP_016859465.1 | ||||
XM_017003977.1 | Intron | XP_016859466.1 | ||||
XM_017003978.1 | Intron | XP_016859467.1 | ||||
XM_017003979.1 | Intron | XP_016859468.1 | ||||
XM_017003980.1 | Intron | XP_016859469.1 |