Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606373 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FMN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FMN2 - formin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305424.1 | 77 | UTR 5 | NP_001292353.1 | |||
NM_020066.4 | 77 | UTR 5 | NP_064450.3 | |||
XM_011544237.2 | 77 | UTR 5 | XP_011542539.1 | |||
XM_017001837.1 | 77 | UTR 5 | XP_016857326.1 | |||
XM_017001838.1 | 77 | UTR 5 | XP_016857327.1 | |||
XM_017001839.1 | 77 | Intron | XP_016857328.1 | |||
XM_017001840.1 | 77 | Intron | XP_016857329.1 | |||
XM_017001841.1 | 77 | Intron | XP_016857330.1 | |||
XM_017001842.1 | 77 | UTR 5 | XP_016857331.1 | |||
XM_017001843.1 | 77 | UTR 5 | XP_016857332.1 |