Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCCCTGCAGAAGTTGCTTAAAA[A/T]TAAGTTACTTGAGAAGAAGCGACTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ATP6V1C2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP6V1C2 - ATPase H+ transporting V1 subunit C2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039362.1 | Intron | NP_001034451.1 | ||||
NM_144583.3 | Intron | NP_653184.2 | ||||
XM_011510339.2 | Intron | XP_011508641.1 | ||||
XM_011510340.2 | Intron | XP_011508642.1 | ||||
XM_011510341.2 | Intron | XP_011508643.1 | ||||
XM_017003745.1 | Intron | XP_016859234.1 |