Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAATTCCACCTCTCAGCAGCCTAG[C/T]CCTGAATCCACACCACAGCAGCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPATA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPATA3 - spermatogenesis associated 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139073.3 | 204 | Silent Mutation | AGC,AGT | S,S 32 | NP_620712.2 | |
XM_005246303.2 | 204 | Silent Mutation | AGC,AGT | S,S 32 | XP_005246360.1 | |
XM_017003363.1 | 204 | Silent Mutation | AGC,AGT | S,S 32 | XP_016858852.1 |
SPATA3-AS1 - SPATA3 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |