Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTGTTCGACACAGTGGAGACCA[C/T]GGGAAAAGACTATTTGTCATCAGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603841 | ||||||||||||||||||||
Literature Links: |
NDUFB5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NDUFB5 - NADH:ubiquinone oxidoreductase subunit B5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199957.1 | 179 | Intron | NP_001186886.1 | |||
NM_001199958.1 | 179 | Silent Mutation | CAC,CAT | H,H 50 | NP_001186887.1 | |
NM_002492.3 | 179 | Silent Mutation | CAC,CAT | H,H 50 | NP_002483.1 |