Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCGGCGGGCGGCAGCTGGCAGCA[C/G]CGCGGCAGGGGCTGCCTTCCCGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607520 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRYL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FRYL - FRY like transcription coactivator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC10A4 - solute carrier family 10 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152679.3 | 166 | Intron | NP_689892.1 |
ZAR1 - zygote arrest 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175619.2 | 166 | Missense Mutation | CAC,CAG | H,Q 42 | NP_783318.1 |