Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGCATTTCTACGACACCGTGAA[G/T]GGCTCCGACTGGCTGGGGGACCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600857 | ||||||||||||||||||||
Literature Links: |
CCDC127 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC127 - coiled-coil domain containing 127 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SDHA - succinate dehydrogenase complex flavoprotein subunit A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001294332.1 | Intron | NP_001281261.1 | ||||
NM_004168.3 | Intron | NP_004159.2 | ||||
XM_005248331.3 | Intron | XP_005248388.1 | ||||
XM_011514072.1 | Intron | XP_011512374.1 | ||||
XM_011514073.1 | Intron | XP_011512375.1 | ||||
XM_017009685.1 | Intron | XP_016865174.1 |