Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCTCTAGTGTGGAGACTTATGC[A/T]GTCCAGATGTTGACCGCAGCCCTGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610774 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNPY3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CNPY3 - canopy FGF signaling regulator 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CNPY3-GNMT - CNPY3-GNMT readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318856.1 | Intron | NP_001305785.1 | ||||
NM_001318857.1 | Intron | NP_001305786.1 | ||||
NM_001318858.1 | Intron | NP_001305787.1 |
Set Membership: |
HapMap |