Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605980 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOD1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NOD1 - nucleotide binding oligomerization domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006092.2 | 3681 | UTR 3 | NP_006083.1 | |||
XM_005249568.1 | 3681 | UTR 3 | XP_005249625.1 | |||
XM_005249572.1 | 3681 | UTR 3 | XP_005249629.1 | |||
XM_005249576.1 | 3681 | UTR 3 | XP_005249633.1 | |||
XM_006715633.2 | 3681 | UTR 3 | XP_006715696.1 | |||
XM_011515079.1 | 3681 | UTR 3 | XP_011513381.1 | |||
XM_011515080.2 | 3681 | UTR 3 | XP_011513382.1 | |||
XM_011515081.2 | 3681 | UTR 3 | XP_011513383.1 | |||
XM_011515083.1 | 3681 | UTR 3 | XP_011513385.1 | |||
XM_011515084.1 | 3681 | UTR 3 | XP_011513386.1 | |||
XM_011515085.1 | 3681 | UTR 3 | XP_011513387.1 | |||
XM_011515087.1 | 3681 | Intron | XP_011513389.1 | |||
XM_011515088.2 | 3681 | Intron | XP_011513390.1 | |||
XM_017011674.1 | 3681 | Intron | XP_016867163.1 | |||
XM_017011675.1 | 3681 | UTR 3 | XP_016867164.1 |
Set Membership: |
HapMap |