Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAATATGAAGTTGCCCTTTCTCA[A/G]TATTTTTCTTTGTACTGCCTGTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613056 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf55-LUC7L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C7orf55-LUC7L2 - C7orf55-LUC7L2 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLRG2 - killer cell lectin like receptor G2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198508.2 | 1565 | Intron | NP_940910.1 | |||
XM_005250311.3 | 1565 | Intron | XP_005250368.1 | |||
XM_011516140.2 | 1565 | Intron | XP_011514442.1 | |||
XM_011516141.2 | 1565 | UTR 3 | XP_011514443.1 |
LOC100129148 - uncharacterized LOC100129148 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LUC7L2 - LUC7 like 2, pre-mRNA splicing factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |