Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGTGCGCATTGCGGGCGGTGAAC[A/G]AGCGCGTGCAGCAGGCTGTGGCGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611605 MIM: 604436 | ||||||||||||||||||||
Literature Links: |
ERLIN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ERLIN2 - ER lipid raft associated 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PROSC - proline synthetase cotranscribed homolog (bacterial) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007198.3 | 971 | Missense Mutation | AAG,GAG | K,E 22 | NP_009129.1 | |
XM_017013001.1 | 971 | Missense Mutation | AAG,GAG | K,E 57 | XP_016868490.1 |