Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607102 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WT1 - Wilms tumor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000378.4 | 2048 | UTR 3 | NP_000369.3 | |||
NM_001198551.1 | 2048 | UTR 3 | NP_001185480.1 | |||
NM_001198552.1 | 2048 | UTR 3 | NP_001185481.1 | |||
NM_024424.3 | 2048 | UTR 3 | NP_077742.2 | |||
NM_024426.4 | 2048 | UTR 3 | NP_077744.3 |