Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGTCAAAACGACATCTATTTCT[C/T]TGCTTGCTTTGTGATCATACCCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176914 | ||||||||||||||||||||
Literature Links: |
PPP1CC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP1CC - protein phosphatase 1 catalytic subunit gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244974.1 | 1235 | Intron | NP_001231903.1 | |||
NM_002710.3 | 1235 | Missense Mutation | AAG,GAG | K,E 322 | NP_002701.1 | |
XM_011538504.2 | 1235 | Intron | XP_011536806.1 | |||
XM_011538505.2 | 1235 | Intron | XP_011536807.1 |