Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACGCACCCGGAGCACAGCTCTGT[C/T]AGGAGCGTGGGTGGGTTGGTGTGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604061 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LACAT1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
LACAT1 - lung adenocarcinoma-associated transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105371907 - uncharacterized LOC105371907 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEPT9 - septin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113491.1 | Intron | NP_001106963.1 | ||||
NM_001113492.1 | Intron | NP_001106964.1 | ||||
NM_001113493.1 | Intron | NP_001106965.1 | ||||
NM_001113494.1 | Intron | NP_001106966.1 | ||||
NM_001113495.1 | Intron | NP_001106967.1 | ||||
NM_001113496.1 | Intron | NP_001106968.1 | ||||
NM_001293695.1 | Intron | NP_001280624.1 | ||||
NM_001293696.1 | Intron | NP_001280625.1 | ||||
NM_001293697.1 | Intron | NP_001280626.1 | ||||
NM_001293698.1 | Intron | NP_001280627.1 | ||||
NM_006640.4 | Intron | NP_006631.2 | ||||
XM_005256962.1 | Intron | XP_005257019.1 | ||||
XM_006721643.2 | Intron | XP_006721706.1 | ||||
XM_006721644.1 | Intron | XP_006721707.1 | ||||
XM_011524204.1 | Intron | XP_011522506.1 | ||||
XM_011524206.1 | Intron | XP_011522508.1 | ||||
XM_011524207.1 | Intron | XP_011522509.1 | ||||
XM_011524208.2 | Intron | XP_011522510.1 | ||||
XM_017024031.1 | Intron | XP_016879520.1 | ||||
XM_017024032.1 | Intron | XP_016879521.1 |
Set Membership: |
HapMap |