Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf47 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C19orf47 - chromosome 19 open reading frame 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256440.1 | 3829 | Intron | NP_001243369.1 | |||
NM_001256441.1 | 3829 | Intron | NP_001243370.1 | |||
XM_005258520.2 | 3829 | Intron | XP_005258577.1 | |||
XM_011526460.2 | 3829 | Intron | XP_011524762.1 | |||
XM_017026291.1 | 3829 | Intron | XP_016881780.1 | |||
XM_017026292.1 | 3829 | UTR 3 | XP_016881781.1 | |||
XM_017026293.1 | 3829 | UTR 3 | XP_016881782.1 | |||
XM_017026294.1 | 3829 | Intron | XP_016881783.1 | |||
XM_017026295.1 | 3829 | Intron | XP_016881784.1 |