Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF233 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF233 - zinc finger protein 233 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207005.1 | 251 | Intron | NP_001193934.1 | |||
NM_181756.2 | 251 | Intron | NP_861421.2 | |||
XM_017026758.1 | 251 | Missense Mutation | CAA,CGA | Q,R 38 | XP_016882247.1 | |
XM_017026759.1 | 251 | Intron | XP_016882248.1 | |||
XM_017026760.1 | 251 | Intron | XP_016882249.1 | |||
XM_017026761.1 | 251 | Missense Mutation | CAA,CGA | Q,R 38 | XP_016882250.1 |