Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCGGGACATGGGGCCCGCCAGCC[A/C]CGCTGCTCGCGGTCTAAGTCGCCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 104775 MIM: 607762 | ||||||||||||||||||||
Literature Links: |
APLP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APLP1 - amyloid beta precursor like protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024807.2 | 209 | Missense Mutation | CAC,CCC | H,P 6 | NP_001019978.1 | |
NM_005166.4 | 209 | Missense Mutation | CAC,CCC | H,P 6 | NP_005157.1 | |
XM_017026737.1 | 209 | Missense Mutation | CAC,CCC | H,P 6 | XP_016882226.1 | |
XM_017026738.1 | 209 | Missense Mutation | CAC,CCC | H,P 6 | XP_016882227.1 |
KIRREL2 - kin of IRRE like 2 (Drosophila) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |