Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGCCAGCACCTGGGCTCTGACGC[C/G]CACTCACTACCTCACCAAGCATGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 180490 | ||||||||||||||||||||
Literature Links: |
MROH8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MROH8 - maestro heat like repeat family member 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPN2 - ribophorin II | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135771.2 | 385 | Missense Mutation | CCC,CGC | P,R 25 | NP_001129243.1 | |
NM_001324299.1 | 385 | Missense Mutation | CCC,CGC | P,R 25 | NP_001311228.1 | |
NM_001324301.1 | 385 | Missense Mutation | CCC,CGC | P,R 25 | NP_001311230.1 | |
NM_001324302.1 | 385 | Missense Mutation | CCC,CGC | P,R 25 | NP_001311231.1 | |
NM_001324303.1 | 385 | Missense Mutation | CCC,CGC | P,R 25 | NP_001311232.1 | |
NM_001324304.1 | 385 | Missense Mutation | CCC,CGC | P,R 25 | NP_001311233.1 | |
NM_001324305.1 | 385 | Missense Mutation | CCC,CGC | P,R 25 | NP_001311234.1 | |
NM_001324306.1 | 385 | UTR 5 | NP_001311235.1 | |||
NM_002951.4 | 385 | Missense Mutation | CCC,CGC | P,R 25 | NP_002942.2 | |
XM_006723851.2 | 385 | Missense Mutation | CCC,CGC | P,R 25 | XP_006723914.1 | |
XM_006723852.2 | 385 | Missense Mutation | CCC,CGC | P,R 25 | XP_006723915.1 |