Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTACTTTATTTCTTCAGGCCTG[A/G]GGTTTTTCGATGACTTCAAATTTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603152 MIM: 606870 | ||||||||||||||||||||
Literature Links: |
ATP5J PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP5J - ATP synthase, H+ transporting, mitochondrial Fo complex subunit F6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003696.1 | 665 | Silent Mutation | CCC,CCT | P,P 106 | NP_001003696.1 | |
NM_001003697.1 | 665 | Silent Mutation | CCC,CCT | P,P 106 | NP_001003697.1 | |
NM_001003701.1 | 665 | Silent Mutation | CCC,CCT | P,P 114 | NP_001003701.1 | |
NM_001003703.1 | 665 | Silent Mutation | CCC,CCT | P,P 106 | NP_001003703.1 | |
NM_001320266.1 | 665 | Silent Mutation | CCC,CCT | P,P 106 | NP_001307195.1 | |
NM_001320267.1 | 665 | Silent Mutation | CCC,CCT | P,P 106 | NP_001307196.1 | |
NM_001685.4 | 665 | Silent Mutation | CCC,CCT | P,P 106 | NP_001676.2 |
JAM2 - junctional adhesion molecule 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |