Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCCTATGCAGAATGGAGAGGAA[G/T]ACCGCCCTTTGGGAGGAGGTGAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300361 | ||||||||||||||||||||
Literature Links: |
BEX3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BEX3 - brain expressed X-linked 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282674.1 | 413 | Missense Mutation | GAC,TAC | D,Y 101 | NP_001269603.1 | |
NM_014380.2 | 413 | Missense Mutation | GAC,TAC | D,Y 21 | NP_055195.1 | |
NM_206915.2 | 413 | Missense Mutation | GAC,TAC | D,Y 21 | NP_996798.1 | |
NM_206917.2 | 413 | Missense Mutation | GAC,TAC | D,Y 11 | NP_996800.1 |