Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCAAGCATCAGTTCTTCCAAAG[A/C]AGCTTAGGTGAAAAACAATTGTTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 309060 | ||||||||||||||||||||
Literature Links: |
LAMP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LAMP2 - lysosomal associated membrane protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122606.1 | 6420 | UTR 3 | NP_001116078.1 | |||
NM_002294.2 | 6420 | UTR 3 | NP_002285.1 | |||
NM_013995.2 | 6420 | Intron | NP_054701.1 |
LOC105373326 - uncharacterized LOC105373326 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |