Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTGGCCACATGGCTAAACCCTG[A/T]CCCATCTCAGAAGCAGAATCTCCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 166490 | ||||||||||||||||||||
Literature Links: |
SPP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPP1 - secreted phosphoprotein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000582.2 | 305 | Missense Mutation | GAC,GTC | D,V 47 | NP_000573.1 | |
NM_001040058.1 | 305 | Missense Mutation | GAC,GTC | D,V 47 | NP_001035147.1 | |
NM_001040060.1 | 305 | Intron | NP_001035149.1 | |||
NM_001251829.1 | 305 | Intron | NP_001238758.1 | |||
NM_001251830.1 | 305 | Missense Mutation | GAC,GTC | D,V 60 | NP_001238759.1 | |
XM_017008564.1 | 305 | Missense Mutation | GAC,GTC | D,V 60 | XP_016864053.1 |