Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGGCAGGTGAACTGGTCGTACCA[G/T]ATGGCCTTGTACATGATCAGCACGC
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 604647 | |||||||||||||||||||||||||||||
Literature Links: |
CALY PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese)
|
|||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CALY - calcyon neuron specific vesicular protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321617.1 | 1620 | UTR 5 | NP_001308546.1 | |||
NM_015722.3 | 1620 | Silent Mutation | NP_056537.1 | |||
XM_005252685.4 | 1620 | Silent Mutation | XP_005252742.2 | |||
XM_011539836.2 | 1620 | Silent Mutation | XP_011538138.1 | |||
XM_011539838.2 | 1620 | Silent Mutation | XP_011538140.1 |