Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTGTAGAACCATCTTCTTCCTC[C/T]TTTTTAAACTTCTATTACTATTTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RAD54L2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAD54L2 - RAD54-like 2 (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322253.1 | Intron | NP_001309182.1 | ||||
NM_001322256.1 | Intron | NP_001309185.1 | ||||
NM_001322257.1 | Intron | NP_001309186.1 | ||||
NM_015106.3 | Intron | NP_055921.2 | ||||
XM_006713062.3 | Intron | XP_006713125.1 | ||||
XM_017005987.1 | Intron | XP_016861476.1 | ||||
XM_017005988.1 | Intron | XP_016861477.1 |