Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTCTTGATGAGCTGGTTCCGCC[G/T]GCGGTGCTCTAGGACACGCTGCAGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 608295 | |||||||||||||||||||||||
Literature Links: |
FAM107A PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
FAM107A - family with sequence similarity 107 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001076778.2 | 932 | Silent Mutation | NP_001070246.1 | |||
NM_001282713.1 | 932 | Silent Mutation | NP_001269642.1 | |||
NM_001282714.1 | 932 | Silent Mutation | NP_001269643.1 | |||
NM_007177.3 | 932 | Silent Mutation | NP_009108.1 |