Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCAGCTTCTCCAGAGCCCACTT[C/T]ACCACCTGGCCAGGGAACGCAGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600047 | ||||||||||||||||||||
Literature Links: |
ABCA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCA2 - ATP binding cassette subfamily A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001606.4 | 6583 | Silent Mutation | GTA,GTG | V,V 2167 | NP_001597.2 | |
NM_212533.2 | 6583 | Silent Mutation | GTA,GTG | V,V 2197 | NP_997698.1 | |
XM_006716996.3 | 6583 | Silent Mutation | GTA,GTG | V,V 2166 | XP_006717059.1 |