Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607725 MIM: 601686 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PARP2 - poly(ADP-ribose) polymerase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEP1 - telomerase associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319035.1 | 7825 | UTR 3 | NP_001305964.1 | |||
NM_007110.4 | 7825 | UTR 3 | NP_009041.2 | |||
XM_005268027.4 | 7825 | UTR 3 | XP_005268084.1 | |||
XM_011537110.2 | 7825 | UTR 3 | XP_011535412.1 |
Set Membership: |
HapMap |