Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGATGCCGTTGGAGGAGTAGTAG[A/T]GGCCCAGGCCCAGGCCGATGAAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605437 MIM: 611029 | ||||||||||||||||||||
Literature Links: |
PRKCH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRKCH - protein kinase C eta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006255.4 | 38 | Intron | NP_006246.2 | |||
XM_011536954.2 | 38 | Intron | XP_011535256.1 | |||
XM_011536955.1 | 38 | Intron | XP_011535257.1 | |||
XM_011536956.1 | 38 | Intron | XP_011535258.1 | |||
XM_011536957.1 | 38 | Intron | XP_011535259.1 | |||
XM_017021458.1 | 38 | UTR 5 | XP_016876947.1 | |||
XM_017021459.1 | 38 | Intron | XP_016876948.1 |
TMEM30B - transmembrane protein 30B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017970.2 | 38 | Missense Mutation | CAC,CTC | H,L 53 | NP_001017970.1 |