Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605437 MIM: 611029 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRKCH PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PRKCH - protein kinase C eta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006255.4 | Intron | NP_006246.2 | ||||
XM_011536954.2 | Intron | XP_011535256.1 | ||||
XM_011536955.1 | Intron | XP_011535257.1 | ||||
XM_011536956.1 | Intron | XP_011535258.1 | ||||
XM_011536957.1 | Intron | XP_011535259.1 | ||||
XM_017021458.1 | Intron | XP_016876947.1 | ||||
XM_017021459.1 | Intron | XP_016876948.1 |
TMEM30B - transmembrane protein 30B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017970.2 | Intron | NP_001017970.1 |
Set Membership: |
HapMap |