Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107400 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERPINA1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SERPINA1 - serpin family A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000295.4 | 2116 | UTR 3 | NP_000286.3 | |||
NM_001002235.2 | 2116 | UTR 3 | NP_001002235.1 | |||
NM_001002236.2 | 2116 | UTR 3 | NP_001002236.1 | |||
NM_001127700.1 | 2116 | UTR 3 | NP_001121172.1 | |||
NM_001127701.1 | 2116 | UTR 3 | NP_001121173.1 | |||
NM_001127702.1 | 2116 | UTR 3 | NP_001121174.1 | |||
NM_001127703.1 | 2116 | UTR 3 | NP_001121175.1 | |||
NM_001127704.1 | 2116 | UTR 3 | NP_001121176.1 | |||
NM_001127705.1 | 2116 | UTR 3 | NP_001121177.1 | |||
NM_001127706.1 | 2116 | UTR 3 | NP_001121178.1 | |||
NM_001127707.1 | 2116 | UTR 3 | NP_001121179.1 | |||
XM_017021370.1 | 2116 | Intron | XP_016876859.1 |
Set Membership: |
HapMap |