Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCAAATTGGAAACATGGCCGTAC[G/T]GTCGCTGTGGGCGGGCCGGCTGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614410 | ||||||||||||||||||||
Literature Links: |
AFAP1L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AFAP1L1 - actin filament associated protein 1 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GRPEL2 - GrpE like 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152407.3 | 137 | Missense Mutation | CGG,CTG | R,L 4 | NP_689620.2 |
GRPEL2-AS1 - GRPEL2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |