Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGGTGCGCCAGGAGGAGAGCGAG[C/T]AGATCAAGACCCTCAACAACAAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 148059 | ||||||||||||||||||||
Literature Links: |
KRT7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRT7 - keratin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005556.3 | 360 | Missense Mutation | CAG,TAG | Q,* 94 | NP_005547.3 | |
XM_011538325.1 | 360 | Missense Mutation | CAG,TAG | Q,* 94 | XP_011536627.1 | |
XM_017019294.1 | 360 | Intron | XP_016874783.1 |
LINC00592 - long intergenic non-protein coding RNA 592 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |