Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCTTTGGCCTAGTGGCCTCCTC[A/C]CTTGTGGATTGGCTGGGTCGCAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MFSD5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MFSD5 - major facilitator superfamily domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170790.1 | 436 | Silent Mutation | TCA,TCC | S,S 204 | NP_001164261.1 | |
NM_032889.4 | 436 | Silent Mutation | TCA,TCC | S,S 97 | NP_116278.3 | |
XM_005269197.1 | 436 | Silent Mutation | TCA,TCC | S,S 204 | XP_005269254.1 | |
XM_005269198.4 | 436 | Silent Mutation | TCA,TCC | S,S 97 | XP_005269255.1 |