Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAAGGTTATGCCCGTACCTGAGG[G/T]AGTGGTATTCCCACCTGATGGGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606092 MIM: 601807 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DNAJC14 - DnaJ heat shock protein family (Hsp40) member C14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMP19 - matrix metallopeptidase 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272101.1 | 1355 | UTR 3 | NP_001259030.1 | |||
NM_002429.5 | 1355 | Missense Mutation | ACC,CCC | T,P 488 | NP_002420.1 | |
XM_006719401.3 | 1355 | Missense Mutation | ACC,CCC | T,P 259 | XP_006719464.1 | |
XM_011538359.1 | 1355 | Missense Mutation | ACC,CCC | T,P 406 | XP_011536661.1 | |
XM_017019308.1 | 1355 | Intron | XP_016874797.1 | |||
XM_017019309.1 | 1355 | Intron | XP_016874798.1 |
TMEM198B - transmembrane protein 198B (pseudogene) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |