Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACATTGGCCACCCAGTCCAGAGT[C/G]AGGTGGTTGGGGTCCTGCAAGTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
KCTD21 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCTD21 - potassium channel tetramerization domain containing 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029859.2 | 638 | Silent Mutation | CTC,CTG | L,L 184 | NP_001025030.1 | |
XM_005273925.3 | 638 | Silent Mutation | CTC,CTG | L,L 253 | XP_005273982.1 | |
XM_005273928.1 | 638 | Silent Mutation | CTC,CTG | L,L 184 | XP_005273985.1 | |
XM_006718516.3 | 638 | Silent Mutation | CTC,CTG | L,L 220 | XP_006718579.1 | |
XM_006718517.2 | 638 | Silent Mutation | CTC,CTG | L,L 184 | XP_006718580.1 | |
XM_006718518.3 | 638 | Silent Mutation | CTC,CTG | L,L 184 | XP_006718581.1 | |
XM_011544955.2 | 638 | Silent Mutation | CTC,CTG | L,L 184 | XP_011543257.1 |
KCTD21-AS1 - KCTD21 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |