Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCGCCCCGTCTCCCTCCCCCCCA[C/T]CACACCGGGCTGTAACCCGAGCCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 176873 MIM: 611141 MIM: 603321 | ||||||||||||||||||||
Literature Links: |
CDK11B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDK11B - cyclin dependent kinase 11B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIB2 - mindbomb E3 ubiquitin protein ligase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMP23B - matrix metallopeptidase 23B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006983.1 | 412 | Intron | NP_008914.1 | |||
XM_017002615.1 | 412 | UTR 5 | XP_016858104.1 | |||
XM_017002616.1 | 412 | UTR 5 | XP_016858105.1 | |||
XM_017002617.1 | 412 | UTR 5 | XP_016858106.1 |