Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
42 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176873 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK11B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK11B - cyclin dependent kinase 11B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291345.1 | 620 | Intron | NP_001278274.1 | |||
NM_001787.2 | 620 | Intron | NP_001778.2 | |||
NM_033486.2 | 620 | Intron | NP_277021.2 | |||
NM_033487.2 | 620 | Intron | NP_277022.1 | |||
NM_033489.2 | 620 | Intron | NP_277024.2 | |||
NM_033490.2 | 620 | Intron | NP_277025.1 | |||
XM_006711065.3 | 620 | UTR 5 | XP_006711128.1 | |||
XM_011542490.2 | 620 | UTR 5 | XP_011540792.1 | |||
XM_011542491.2 | 620 | Intron | XP_011540793.1 | |||
XM_011542492.2 | 620 | Intron | XP_011540794.1 | |||
XM_011542493.2 | 620 | UTR 5 | XP_011540795.1 | |||
XM_011542494.2 | 620 | UTR 5 | XP_011540796.1 | |||
XM_011542495.2 | 620 | UTR 5 | XP_011540797.1 | |||
XM_011542496.2 | 620 | Intron | XP_011540798.1 | |||
XM_017002925.1 | 620 | UTR 5 | XP_016858414.1 | |||
XM_017002926.1 | 620 | UTR 5 | XP_016858415.1 | |||
XM_017002927.1 | 620 | UTR 5 | XP_016858416.1 | |||
XM_017002928.1 | 620 | UTR 5 | XP_016858417.1 |
SLC35E2B - solute carrier family 35 member E2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110781.2 | 620 | Intron | NP_001104251.1 | |||
NM_001290264.1 | 620 | Intron | NP_001277193.1 |