Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGTCAGGTTGATTACAGATATA[C/G]CAAATCTATTTTTCCTAGGGTAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602516 | ||||||||||||||||||||
Literature Links: |
RGS4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RGS4 - regulator of G-protein signaling 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102445.2 | 148 | UTR 5 | NP_001095915.1 | |||
NM_001113380.1 | 148 | Intron | NP_001106851.1 | |||
NM_001113381.1 | 148 | Intron | NP_001106852.1 | |||
NM_005613.5 | 148 | Intron | NP_005604.1 |